Learn More
Abcam Human LAIR1 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Product Code. 32113386 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32113386 10 x 96 Tests 96 tests
1 options
Product Code. 32113386 Supplier Abcam Supplier No. ab30509210x96Tests
 Restricted Item
Forventet forsendelse: 27-11-2026
Antibody, suitable for sELISA.
  • Applications: sELISA
  • Species Reactivity: Human
Specifications
Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols LAIR1
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human.
Target Species Human
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv sELISA