Learn More
Abcam Human IL-6 Matched Antibody Pair Kit, 5 x 96 Tests
Product Code. 32118561 Shop All Abcam Products
Change view
Click to view available options
Quantity:
5 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32118561 5 x 96 Tests 96 tests
1 options
Product Code. 32118561 Supplier Abcam Supplier No. ab2468385x96Tests
 Restricted Item
Forventet forsendelse: 26-11-2026
Antibody, suitable for ELISA, with 4 citations.
  • Applications: ELISA
  • Species Reactivity: Human
Specifications
Applications ELISA
Conjugate Unconjugated
Gene Symbols IL6
Quantity 5 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human.
Target Species Human
Product Type Secondary Antibody
Detection Method Colorimetric
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv ELISA