Learn More
Abcam Human AKT1 (phospho S473) + AKT3 (phospho S472) Antibody Pair - BSA and Azide free, 10 x 96 Tests
Product Code. 32112529 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32112529 10 x 96 Tests 96 tests
1 options
Product Code. 32112529 Supplier Abcam Supplier No. ab25386710x96Tests
 Restricted Item
Forventet forsendelse: 26-11-2026
Antibody, suitable for sELISA.
  • Applications: sELISA
  • Species Reactivity: Human
Specifications
Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols AKT1
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human.
Target Species Human
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Hu
Target Modification State phospho S473
Validated Application abrv sELISA