Få mere at vide
Abcam Anti-IgG antibody [A set of murine monoclonal antibodies], 100 ug
Artikelnummer. 32053157 Shop alle Abcam produkter
Skift visning
Mængde:
100 μg
Artikelnummer. Mængde
32053157 100 μg
1 options
Artikelnummer. 32053157 Leverandør Abcam Leverandørnr. ab210935100ug
 Begrænset vare
Forventet forsendelse: 12-11-2026
Mouse Monoclonal antibody, suitable for ELISA, WB, with 3 citations.
  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: ELISA, WB
  • Species Reactivity: Rat
  • Storage Buffer: Preservative: 0.09% Sodium azide, Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification Protein A
The product is subject to the following: Abcam Restricted Use Statement
Tekniske data
Antigen IgG
Applikationer ELISA, Western Blot
Klassifikation Monoclonal
Klon A set of murine monoclonal antibodies
Konjugeret Unconjugated
Formulering Preservative: 0.09% Sodium azide, Constituents: PBS
Gene IgG
Gensymboler IgG
Værtsarter Mouse
Oprensningsmetode Affinity purification Protein A
Mængde 100 μg
Regulatorisk status RUO
Hele molekyle Yes
Primær eller sekundær Primary
Emballage Vial
Testspecificitet This product reacts with Rat.
Målarter Rat
Produkttype Monoclonal Antibody (IgG)
Form Liquid
Isotype IgG
Vært abrv Ms
Forsendelsestilstand Blue Ice
Reaktivitet abrv Rt
Target Gene IgG
Målindgang IgG
Målmolekyle IgG, Rat
Teknik ELISA, WB
Valideret ansøgning abrv ELISA, WB