Learn More
Bellingham Stanley REFRACTOMETER 60/ED ABBE HIGH ACCURACY 230V 50/60Hz 5W +/-0.00004 RI range of 1.30-1.74 approx Bellingham and Stanley, 1/Cs, MA: TBD
Brand: Bellingham Stanley 10-04
REFRACTOMETER 60/ED ABBE HIGH ACCURACY 230V 50/60Hz 5W +/-0.00004 RI range of 1.30-1.74 approx Bellingham and Stanley, 1/Cs, MA: TBD
Additional Details : Weight : 0.01000kg