Få mere at vide
Abacavir-d4 (cyclopropyl-2,2,3,3-d4), CDN
Artikelnummer. 30326582
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326582 0.01 g
30323557 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326582 Leverandør CDN Leverandørnr. CDND80390.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Abacavir-D4 (cyclopropyl-2,2,3,3-D4)
CAS 1260619-56-4
Molekylær formel C14 D4 H14 N6 O
Mængde 0.01 g
Synonym Abacavir-D4, 2-Cyclopentene-1-methanol, 4-[2-amino-6-(cyclopropyl-2,2,3,3-d4-amino)-9 H-purin-9-yl]-, (1 S,4 R)- (ACI), (1 S,4 R)-4-[2-Amino-6-(cyclopropyl-2,2,3,3-d4-amino)-9 H-purin-9-yl]-2-cyclopentene-1-methanol (ACI), Abacavir-D4 (cyclopropyl-2,2,3,3-D4), Abacavir-d4
InChI formel InChI=1 S/C14H18N6O/c15-14-18-12(17-9-2-3-9)11-13(19-14)20(7-16-11)10-4-1-8(5-10)6-21/h1,4,7-10,21 H,2-3,5-6H2,(H3,15,17,18,19)/t8-,10+/m1/s1/i2D2,3D2
SMIL [2 H]C1([2 H])C(Nc2nc(N)nc3c2ncn3[C@@H]4 C[C@H](CO)C=C4)C1([2 H])[2 H]
IUPAC navn [(1 S,4 R)-4-[2-amino-6-[(2,2,3,3-tetradeuteriocyclopropyl)amino]purin-9-yl]cyclopent-2-en-1-yl]methanol
Molekylvægt (g/mol) 290.357
Formel vægt 290.1793 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity