Få mere at vide
Invitrogen™ AAV Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14106
Beskrivelse
PA1-4106 detects adeno-associated virus. PA1-4106 has been successfully used in Western blot procedures. The PA1-4106 immunogen is a mix of synthetic peptides corresponding to residues T(75) P W G Y F D F N R F H C(86) of major coat protein VP3 [AAV 2], residues E(278) G G T N L L Q F P H V(289) of capsid protein VP1 [AAV 6], and residues 277-288 of capsid AAV 3.
Adeno-associated virus (AAV) is a small virus which infects humans and some other primate species. AAV is not currently known to cause disease. The virus causes a very mild immune response, lending further support to its apparent lack of pathogenicity.
Tekniske data
| AAV | |
| Polyclonal | |
| Unconjugated | |
| AAV; AdV; HADV; Lentivirus | |
| Synthetic peptides corresponding to residues 75-86 of major coat protein VP3 [AAV 2], 278-289 of capsid protein VP1 [AAV 6], and 277-288 of capsid AAV 3. | |
| 100 μg | |
| Primary | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.