Få mere at vide
Tocris Bioscience™ A66
Potent and selective PI 3-kinase p110α inhibitor
Brand: Tocris Bioscience™ 5595/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
A66 is a potent and selective PI 3-kinase p110α inhibitor (IC50 = 32 nM). Exhibits >100-fold selectivity for p110α over other PI 3-kinase isoforms. Inhibits Akt signaling and tumor growth in SK-OV-3 xenografts in mice.Tekniske data
| (2S)-N1-[2-(1,1-Dimethylethyl)-4′-methyl[4,5′-bithiazol]-2′-yl]-1,2-pyrrolidinedicarboxamide | |
| 1166227-08-2 | |
| 10 mg |