Få mere at vide
Invitrogen™ A2B5 Monoclonal Antibody (105)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA190445
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
A2B5 binds to neural tissue such as retina, brain, spinal cord, and dorsal root ganglia. It is also able to bind plasma membranes of human, rat, chicken and mouse islet cells and can be used in cell sorting.
Tekniske data
| A2B5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Mouse | |
| IgM | |
| RUO | |
| Human, Mouse, Rat, Chicken | |
| Antibody | |
| IgM |
| Immunocytochemistry | |
| 105 | |
| Unconjugated | |
| A2B5 marker | |
| Embryonic chicken retinal cells. | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |