Learn More
Moldex™ A2 8500 Gas Filter For 8000 Series Respiratory Cartridges
For protection against organic + inorganic gases and vapours
Brand: Moldex™ 850001
Additional Details : Weight : 1.00800kg
Description
Moldex 8000 Series A2 gas filter (8500). A2 gas filter for the 8000 Series half mask. For protection against organic + inorganic gases and vapours, boiling point >65°C. 10 x WEL or 5, 000ppm whichever is lower.Specifications
A2 8500 | |
Series Filter | |
10/Pk. |