Learn More
Moldex™ A1B1E1K1 Gas Filter For 8000 Series Respiratory Cartridges
For protection against organic + inorganic gases and vapours
Brand:  Moldex™ 890001
Additional Details : Weight : 1.05280kg
Product Code. 12372309
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Moldex 8000 Series ABEK1 gas filter (8900). ABEK1 gas filter for the 8000 Series half mask. For protection against organic + inorganic gases and vapours, boiling point >65°C, + ammonia and acid gases. 10 x WEL or 1,000ppm whichever is lower.
Specifications
Specifications
A1B1E1K1
Gas Filter
For Series 8000 Respiratory Filter
10/Pk.