Få mere at vide
(-)-a-Pinene-10,10,10-d3, CDN
Artikelnummer. 30323781
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30323781 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323781 Leverandør CDN Leverandørnr. CDND80250.01G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1 S)-(-)-alpha-Pinene-D3
CAS 1903007-82-8
Molekylær formel C10 D3 H13
Mængde 0.01 g
Synonym (-)-a-Pinene-10,10,10-d3, (1 S)-(-)-α-Pinene-D3, (1 S,5 S)-6,6-Dimethyl-2-(methyl-d3)bicyclo[3.1.1 ]hept-2-ene, (-)-α-Pinene-10,10,10-D3
InChI formel InChI=1 S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h4,8-9 H,5-6H2,1-3H3/t8-,9-/m0/s1/i1D3
SMIL [2 H]C([2 H])([2 H])C1=CC[C@H]2 C[C@@H]1C2(C)C
IUPAC navn (1 S,5 S)-6,6-dimethyl-2-(trideuteriomethyl)bicyclo[3.1.1 ]hept-2-ene
Molekylvægt (g/mol) 139.252
Formel vægt 139.144 g/mol
Procent renhed 99 atom % D, min 97% Chemical Purity