Få mere at vide
a-(N-Methyl-N-ethylamino)butyrophenone-d3 Hydrochloride, TRC
Artikelnummer. 30473482
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30473482 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473482 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30485225MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha-(N-Methyl-N-ethylamino)butyrophenone-d3 Hydrochloride
CAS 1794737-35-1
Molekylær formel C13H17D3ClNO
Mængde 25 mg
Synonym 2-[N-(Methyl-d3)-N-ethylamino]-1-phenyl-1-butanone Hydrochloride
InChI formel InChI=1S/C13H19NO.ClH/c1-4-12(14(3)5-2)13(15)11-9-7-6-8-10-11;/h6-10,12H,4-5H2,1-3H3;1H/i3D3;
SMIL [2H]C([2H])([2H])N(CC)C(CC)C(=O)C1=CC=CC=C1.Cl
IUPAC navn 2-[ethyl(trideuteriomethyl)amino]-1-phenylbutan-1-one;hydrochloride
Molekylvægt (g/mol) 244.78
Formel vægt 244.1422