Få mere at vide
Tocris Bioscience™ A 971432
Potent and selective S1P5 agonist
Brand: Tocris Bioscience™ 5766/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
A 971432 is a potent and selective S1P5 agonist (EC50 values are 4.1 and 5.7 nM in cAMP and GTPγS assays, respectively). Exhibits 60-fold selectivity over S1P1, and >1,600-fold selectivity over S1P2-4, and a panel of 129 kinases and 79 receptors. Improves blood-brain barrier integrity in vitro and reverses age-related cognitive decline in vivo. Orally available.Tekniske data
| 1-[[4-[(3,4-Dichlorophenyl)methoxy]phenyl]methyl]-3-azetidinecarboxylic acid | |
| 1240308-45-5 | |
| 10 mg |