Få mere at vide
Tocris Bioscience™ A 779
Selective Mas receptor (Ang-(1-7) receptor) antagonist
Brand: Tocris Bioscience™ 5937/5
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
A 779 is a selective Mas receptor (Ang-(1-7) receptor) antagonist. Exhibits no significant affinity for AT1 or AT2 receptors at a concentration of 1 μM. Inhibits antidiuretic effect of Ang-(1-7) in water-loaded rats. Also attenuates Monocrotaline-induced pulmonary fibrosis in rats.Tekniske data
| 0.95 | |
| Store at -20°C | |
| Observed M.W.: 872.98 | |
| C39H60N12O11 |
| 159432-28-7 | |
| Other Angiotensin Receptors | |
| 5 mg |