missing translation for 'onlineSavingsMsg'
Få mere at vide

A 769662, TRC

Artikelnummer. 30327415
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327415 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30327415 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1012055MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale A 769662
CAS 844499-71-4
Molekylær formel C20H12N2O3S
Mængde 5 mg
Synonym 4-Hydroxy-3-(2'-hydroxy-1,1'-biphenyl-4-yl)-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carbonitrile, 6,7-Dihydro-4-hydroxy-3-(2'-hydroxy[1,1'-biphenyl]-4-yl)-6-oxo-thieno[2,3-b]pyridine-5-carbonitrile
InChI formel InChI=1S/C20H12N2O3S/c21-9-14-18(24)17-15(10-26-20(17)22-19(14)25)12-7-5-11(6-8-12)13-3-1-2-4-16(13)23/h1-8,10,23H,(H2,22,24,25)
SMIL [Na+].CCC(=O)O[C@H](C(C)C)OP(CCCCc1ccccc1)(=O)CC(=O)N2C([C@@H](=O)[O-])C[C@@H](C3CCCCC3)C2
IUPAC navn 4-hydroxy-3-[4-(2-hydroxyphenyl)phenyl]-6-oxo-7H-thieno[2,3-b]pyridine-5-carbonitrile
Molekylvægt (g/mol) 360.39
Formel vægt 360.0569
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.