Learn More
Tygon A-60-G Chemical Dispensing Tubing
Specially formulated tubing offers excellent resistance to inorganic fluids
Brand: Tygon 50106-68
This item is not returnable.
View return policy
Description
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. Outstanding resistance to heat, UV, and ozone. Use in temperatures ranging from -60°C to 135°C (-75°F to 275°F). High flexural fatigue strength—ideal for use in peristaltic pumps. Heat sealable—no fittings needed to join tubingSpecifications
1 in. | |
1-1/4 in. | |
Black |
Tygon A-60-G | |
50 ft. |
Product Content Correction
Product Title
Tygon A-60-G Chemical Dispensing Tubing > 1x1-1/4 in.
Spot an opportunity for improvement?Share a Content Correction