Få mere at vide
Tygon A-60-G Chemical Dispensing Tubing
Specielt formuleret slange giver fremragende modstandsdygtighed over for uorganiske væsker
Brand:  Tygon 50106-64
 Se flere versioner af dette produkt
Produktkode 30053538
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. Outstanding resistance to heat, UV, and ozone. Use in temperatures ranging from -60°C to 135°C (-75°F to 275°F). High flexural fatigue strength—ideal for use in peristaltic pumps. Heat sealable—no fittings needed to join tubing
Tekniske data
Tekniske data
1/2 in.
11/16 in.
Black
Tygon A-60-G
50 ft.
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
One moment while we fetch your results.
Certifikater
Kampagner
Kampagner
Korrektion af produktindhold
Produkttitel
Tygon A-60-G Chemical Dispensing Tubing > 11/16 in.,1/2 in.