Learn More
Tygon A-60-G Chemical Dispensing Tubing
Specially formulated tubing offers excellent resistance to inorganic fluids
Brand:  Tygon 50106-50
 View more versions of this product
Product Code. 30053394
  • 456.79 DKK / 15m
Only null left
Estimated Shipment: 06-10-2025
Add to basket
This item is not returnable. View return policy
Description
Description
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. Outstanding resistance to heat, UV, and ozone. Use in temperatures ranging from -60°C to 135°C (-75°F to 275°F). High flexural fatigue strength—ideal for use in peristaltic pumps. Heat sealable—no fittings needed to join tubing
Specifications
Specifications
1/16 in.
3/16 in.
Black
Tygon A-60-G
50 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tygon A-60-G Chemical Dispensing Tubing > 3/16 in.,1/16 in.