Få mere at vide
A 485, Tocris Bioscience™
GREENER_CHOICE
Histone Acetyltransferase Inhibitors
Brand:  Tocris Bioscience 6387/5
Encompass
Produktkode 15997930
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
A 485 is a potent and selective p300/CREB-binding protein (CBP) HAT domain inhibitor (IC50 values are 2.6 and 9.8 nM for the CBP-bromodomain HAT-C/H3 (BHC) and p300-BHC domains, respectively), which displays > 1000-fold selectivity over closely related HATs. A 485 suppresses proliferation in several hematological malignancies and AR+ prostate cancer cell lines in vitro, and also inhibits tumor growth in a castration-resistant prostate cancer xenograft model. A 485 is orally bioavailable.
To request the negative control for A 485, please fill out the A 486 request form on the SGC website.
Tekniske data
Tekniske data
1889279-16-6
(1R)-N-[(4-Fluorophenyl)methyl]-2,3-dihydro-5-[[(methylamino)carbonyl]amino]-2′,4′-dioxo-N-[(1S)-2,2,2-trifluoro-1-methylethyl]spiro[1H-indene-1,5′-oxazolidine]-3′-acetamide
5 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner