Få mere at vide
Tocris Bioscience™ A 438079 hydrochloride
GREENER_CHOICE
Artikelnummer. 18014842
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18014842 10 mg
18015682 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18014842 Leverandør Tocris Bioscience™ Leverandørnr. 2972/10
Competitive P2X7 antagonist
A 438079 hydrochloride is a competitive P2X7 receptor antagonist (pIC50 = 6.9 for the inhibition of Ca2+ influx in the human recombinant P2X7 cell line). Devoid of activity at other P2 receptors (IC50 > 10 μM). Possesses antinociceptive activity in models of neuropathic pain in vivo. Inhibits methamphetamine-induced microglial migration and phagocytosis in vitro.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 899431-18-6
Molekylær formel C13H10Cl3N5
Tekniske data
Kemisk navn eller materiale A 438079 hydrochloride
Mængde 10 mg
Formel vægt 342.61
Renhed >98%
Molekylvægt (g/mol) 342.608