Få mere at vide
Tocris Bioscience™ A 425619
GREENER_CHOICE
Potent TRPV1 antagonist
Brand:  Tocris Bioscience™ 5781/10
Encompass
Produktkode 15717116
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
A 425619 is a potent TRPV1 antagonist (IC50 = 3-9 nM). Blocks capsaicin-evoked currents in rat DRG neurons and inhibits TRPV1-activation by anandamide (Cat. No. 1339) and N-arachidonoyl-dopamine. Reduces pain in inflammatory, postoperative and osteoarthritic in vivo pain models.
Tekniske data
Tekniske data
N-5-Isoquinolinyl-N′-[[(4-(trifluoromethyl)phenyl]methyl]urea
581809-67-8
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner