Få mere at vide
Tocris Bioscience™ A 425619
Potent TRPV1 antagonist
Brand: Tocris Bioscience™ 5781/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
A 425619 is a potent TRPV1 antagonist (IC50 = 3-9 nM). Blocks capsaicin-evoked currents in rat DRG neurons and inhibits TRPV1-activation by anandamide (Cat. No. 1339) and N-arachidonoyl-dopamine. Reduces pain in inflammatory, postoperative and osteoarthritic in vivo pain models.Tekniske data
| N-5-Isoquinolinyl-N′-[[(4-(trifluoromethyl)phenyl]methyl]urea | |
| 581809-67-8 | |
| 50 mg |