missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ A 410099.1, amine
GREENER_CHOICE

Artikelnummer. 18736806
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18736806 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18736806 Leverandør Tocris Bioscience™ Leverandørnr. 6471/100

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Functionalized IAP ligand for PROTACs

A 410099.1, amine is a functionalized IAP ligand for PROTAC® research and development. Boc protected A 410099.1 (Cat.No. 6470). Supplied with an amine functional handle for ready conjugation to a linker/target protein ligand. NanoBRET assays show EC50 values of 4.6, 9.2 and 15.6 nM for cIAP1, cIAP2 and XIAP proteins, respectively. Part of a range of functionalized tool molecules for PROTAC R&D. Please contact us for SD files of our available Degrader Building Blocks. PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale tert-Butyl [(S)-1-[[(S)-2-[(2S,4S)-4-amino-2-[[(R)-1,2,3,4-tetrahydronaphthalen-1-yl]carbamoyl]pyrrolidin-1-yl]-1-cyclohexyl-2-oxoethyl]amino]-1-oxopropan-2-yl](methyl)carbamate hydrochloride
CAS 2374122-37-7
Mængde 100 mg
Molekylær formel C32H49N5O5.HCl
Renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.