missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ A 410099.1 amide-PEG4-amine

Description
A 410099.1 amide-PEG4-amine is a functionalized IAP ligand for PROTAC™ research and development; incorporates an IAP ligand plus an amide-PEG4 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Kemisk navn eller materiale | tert-Butyl (-1-((-2-((2 S,4 S)-4-(14-amino-3,6,9,12-tetraoxatetradecanamido)-2-(((R)-1,2,3,4-tetrahydronaphthalen-1-yl)carbamoyl)pyrrolidin-1-yl)-1-cyclohexyl-2-oxoethyl)amino)-1-oxopropan-2-yl)(methyl)carbamate |
| CAS | 2446474-10-6 |
| Mængde | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylær formel | C42H68N6O10 |
| Renhed | 0.95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?