Få mere at vide
A-1331852, TRC
Artikelnummer. 30179672
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179672 25 mg
30179647 100 mg
30179115 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179672 Leverandør Toronto Research Chemicals Leverandørnr. TRCA10090025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1430844-80-6
Molekylær formel C38H38N6O3S
Synonym 6-[8-[(2-Benzothiazolylamino)carbonyl]-3,4-dihydro-2(1H)-isoquinolinyl]-3-[5-methyl-1-(tricyclo[3.3.1.13,7]dec-1-ylmethyl)-1H-pyrazol-4-yl]-2-pyridinecarboxylic Acid
Tekniske data
Kemisk navn eller materiale A-1331852
Mængde 25 mg
Forsendelsestilstand Room Temperature
Formel vægt 658.2726
MDL nummer MFCD30532662
Molekylvægt (g/mol) 658.81, 658.82
InChI nøgle QCQQONWEDCOTBV-UHFFFAOYSA-N
SMIL [H]C12CC3([H])CC([H])(C1)CC(CN1N=CC(=C1C)C1=CC=C(N=C1C(O)=O)N1CCC4=C(C1)C(=CC=C4)C(=O)NC1=NC4=CC=CC=C4S1)(C2)C3
IUPAC navn 3-{1-[(adamantan-1-yl)methyl]-5-methyl-1H-pyrazol-4-yl}-6-{8-[(1,3-benzothiazol-2-yl)carbamoyl]-1,2,3,4-tetrahydroisoquinolin-2-yl}pyridine-2-carboxylic acid