Få mere at vide
8-Cyclopentyl-1,3-dimethylxanthine, Thermo Scientific™
A selective A1 adenosine receptor antagonist
Brand: Thermo Scientific Alfa Aesar J61565.MC
| Mængde | 100mg |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
| 8-Cyclopentyl-1,3-dimethylxanthine | |
| C12H16N4O2 | |
| 100mg | |
| Soluble in 0;1M NaOH; Slightly soluble in water; Also soluble in DMSO at 16mg/ml at 60°C. | |
| CN1C2=C(C(=O)N(C1=O)C)NC(=N2)C3CCCC3 | |
| 248.286 | |
| 248.28 |
| 35873-49-5 | |
| MFCD00055116 | |
| 8-cyclopentyltheophylline, 8-cyclopentyl-1,3-dimethylxanthine, 8-cyclopentyl theophylline, 1,3-dimethyl-8-cyclopentylxanthine, xanthine, 8-cyclopentyl-1,3-dimethyl, theophylline, 8-cyclopentyl, 8-cyclopentyl-1,3-dimethyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6-dione, 8-cyclopentyl-3,9-dihydro-1,3-dimethyl, spectrum_000351, lopac-c-102 | |
| SCVHFRLUNIOSGI-UHFFFAOYSA-N | |
| 8-cyclopentyl-1,3-dimethyl-7H-purine-2,6-dione | |
| 1917 |