Få mere at vide
Brompheniramine maleate, Thermo Scientific™
An antihistamine and also an inhibitor of serotonin reuptake
Brand: Thermo Scientific Alfa Aesar J63922.MB
| Mængde | 25mg |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
| Brompheniramine maleate | |
| C20H23BrN2O4 | |
| 25mg | |
| brompheniramine maleate, brompheniramine hydrogen maleate, dimetane, rolabromophen, dimotane, drauxin, gammistin, nagemid, spentane, allent | |
| OC(=O)\C=C/C(O)=O.CN(C)CCC(C1=CC=C(Br)C=C1)C1=CC=CC=N1 | |
| 435.32 | |
| CHEBI:3184 |
| 980-71-2 | |
| MFCD00057367 | |
| 14,1443 | |
| SRGKFVAASLQVBO-BTJKTKAUNA-N | |
| (2Z)-but-2-enedioic acid; [3-(4-bromophenyl)-3-(pyridin-2-yl)propyl]dimethylamine | |
| 5281067 | |
| 319.24 |