missing translation for 'onlineSavingsMsg'
Få mere at vide

Cerulenin, 98%, Thermo Scientific™

Has antibiotic and antifungal properties; inhibits fatty acid and steroid biosynthesis

Brand:  Thermo Scientific Alfa Aesar J64538.MA

Mængde 10mg

Produktkode 15426449

  • ONLINE EXCLUSIVE PRICE
    2611.98 DKK
    2349.65 DKK / 10mg
    Spare 262.33 DKK
    10% Slukket

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cerulenin is used as an antifungal antibiotic that inhibits sterol and fatty acids biosynthesis. Cerulenin induces apoptosis in tumor cell lines. It can be a useful biochemical tool with applications for the study of metabolism and function of fatty acids in the bacterial cell membrane and in yeast. An extensive review including the properties, uses, mechanism of action and structure has been reported.

Solubility
Soluble in acetone at 20mg/ml. Also soluble in ethanol, benzene, chloroform, or ethyl acetate.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers..
TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

Cerulenin
White to Yellow
C12H17NO3
4140423
(2R,3S,E,E)-2,3-Epoxy-4-oxo-7,10-dodecadienamide
GVEZIHKRYBHEFX-NQQPLRFYSA-N
(2R,3S)-3-[(4E,7E)-nona-4,7-dienoyl]oxirane-2-carboxamide
5282054
223.27
17397-89-6
10mg
MFCD00077686
14,2004
Soluble in acetone at 20mg/ml; Also soluble in ethanol,benzene,chloroform,or ethyl acetate.
CC=CCC=CCCC(=O)C1C(O1)C(=O)N
223.272
CHEBI:171741
98%
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.