Få mere at vide
Tacrine hydrochloride, Thermo Scientific™
A potent centrally acting anticholinesterase
Brand: Thermo Scientific Alfa Aesar J60070.06
| Mængde | 5g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in aqueous buffers
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Tekniske data
| Tacrine hydrochloride | |
| C13H15ClN2 | |
| 5g | |
| 14,9024 | |
| Soluble in aqueous buffers | |
| [H+].[Cl-].NC1=C2C=CC=CC2=NC2=C1CCCC2 | |
| 234.73 | |
| 234.73 (Anhydrous) |
| 1684-40-8 | |
| MFCD00150067 | |
| UN2811 | |
| tacrine hydrochloride, 1,2,3,4-tetrahydroacridin-9-amine hydrochloride, tacrine hcl, hydroaminacrine, tenakrin, 9-amino-1,2,3,4-tetrahydroacridine hydrochloride, tacrine.hcl, unii-4966rng0bu, 1,2,3,4-tetrahydro-9-acridinamine hydrochloride, ccris 7250 | |
| ZUFVXZVXEJHHBN-UHFFFAOYSA-N | |
| hydrogen 1,2,3,4-tetrahydroacridin-9-amine chloride | |
| 2723754 | |
| ≥98% |