Få mere at vide
Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III), Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 088481.01
| Mængde | 0.25g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
| Tris (trifluoromethylhydroxymethylene-d-camphorato)europium (III) | |
| C36H45EuF9O6 | |
| 0.25g | |
| eu tfc 3, eu facam 3, europium tris 3-trifluoromethylhydroxymethylene---camphorate, europium tris 3-trifluoromethylhydroxymethylene-+-camphorate, tris 3-trifluoromethylhydroxymethylene-d-camphorato europium iii, tris 3-trifluoroacetyl-d-camphorato europium iii, europium iii tris 3-trifluoromethylhydroxymethylene-d-camphorate | |
| YXJPWWGLCOPUGI-UFRADDTFSA-N | |
| europium;(2E)-4,7,7-trimethyl-2-(2,2,2-trifluoro-1-hydroxyethylidene)bicyclo[2.2.1]heptan-3-one | |
| 71310195 |
| 34830-11-0 | |
| MFCD00074803 | |
| Moisture sensitive | |
| Soluble in chloroform. Insoluble in water. | |
| CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.[Eu] | |
| 896.7 | |
| 893.68 |