Få mere at vide
Gilson™ Maintenance Kit for Pipetman™
Manufactured by Gilson™. Gilson™ Maintenance Kit for Pipetman™ are guaranteed to deliver value and assure reliability and precision.
Brand: Gilson™ F144495
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
- Maintenance kit
Tekniske data
| For Pipetman™ P20/F2-F20 | |
| 1 Tip-holder, 3 piston seals and 3 O-rings |