Få mere at vide
(9Z,11E)-Methyl Ester 9,11-Octadecadienoate, TRC
Artikelnummer. 30009119
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30009119 10 mg -
30009971 25 mg -
30009532 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009119 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32530510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13058-52-1
Molekylær formel C19 H34 O2
Molekylvægt (g/mol) 294.47
Synonym Methyl 9-cis-11-trans-octadecadienoate
IUPAC navn methyl (9Z,11E)-octadeca-9,11-dienoate
SMIL CCCCCC\C=C\C=C/CCCCCCCC(=O)OC
Tekniske data
Kemisk navn eller materiale (9Z,11E)-Linoleic acid methyl ester
Mængde 10 mg
InChI formel InChI=1S/C19H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h8-11H,3-7,12-18H2,1-2H3/b9-8+,11-10-
Formel vægt 294.2559