Få mere at vide
9alpha-Fluoro-11beta-hydroxy-16-beta-methyl-3-oxoandrosta-1,4-diene-17(R)-spiro-2’-[4’-chloro-5’-ethylfuran-3’(2’H)-one, TRC
Artikelnummer. 30009107
Skift visning
Mængde:
25 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009107 25 mg
30003243 500 mg
30004690 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009107 Leverandør Toronto Research Chemicals Leverandørnr. TRCF59180025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1486466-31-2
Molekylær formel C25 H30 Cl F O4
Molekylvægt (g/mol) 448.95
Synonym (17R)-4'-Chloro-5'-ethyl-9-fluoro-11β-hydroxy-16β-methylspiro[androsta-1,4-diene-17,2'(3'H)-furan]-3,3'-dione, Clobetasol Propionate 17α-spiro Compound, 9α-Fluoro-11β-hydroxy-16β-methyl-3-oxoandrosta-1,4-diene-17(R)-spiro-2'-[4'-chloro-5'-ethylfuran-3'(2'H)-one
IUPAC navn (8S,9R,10S,11S,13S,14S,16S,17R)-4'-chloro-5'-ethyl-9-fluoro-11-hydroxy-10,13,16-trimethylspiro[6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17,2'-furan]-3,3'-dione
SMIL CCC1=C(Cl)C(=O)[C@@]2(O1)[C@@H](C)C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C
Tekniske data
Kemisk navn eller materiale Clobetasol Propionate 17alpha-spiro Compound
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C25H30ClFO4/c1-5-18-20(26)21(30)25(31-18)13(2)10-17-16-7-6-14-11-15(28)8-9-22(14,3)24(16,27)19(29)12-23(17,25)4/h8-9,11,13,16-17,19,29H,5-7,10,12H2,1-4H3/t13-,16-,17-,19-,22-,23-,24-,25-/m0/s1
Formel vægt 448.1817
Procent renhed >95%