Beskrivelse
- Mobile external battery for use with the uvex protection flash line
- Bright, reflective stripes increase worker safety in poorly lit working conditions
- For use with vests (17255, 89909, and 8971) and softshell jacket (89910)
- Lasts up to 8 hours on a single charge
Tekniske data
Tekniske data
| Genopladelig | Yes |
| Til brug med (udstyr) | Active Flash Line Vests (17255, 89909, 89718), and Softshell Jacket (89910) |
| Dimensioner (L x B x H) | 113 x 58 x 12 mm |
| Vægt (metrisk) | 100 g |