Learn More
Uvex™ 9790 Chinstraps
Product Code. 17289191 Shop All Uvex Products
Change view
Click to view available options
Material:
Elastic
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
17289191 Elastic Each
17299191 - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17289191 Supplier Uvex™ Supplier No. 9790031
  • ONLINE EXCLUSIVE PRICE
    36.10 DKK
    33.43 DKK / Pakke med 1
    Spare 2.67 DKK
    7% Slukket
Forventet forsendelse: 11-12-2026
Chinstraps for use with Uvex™ Safety Helmets available in various materials.
  • For use with Uvex™ Safety Helmets
TRUSTED_SUSTAINABILITY
Specifications
Color Gray, Green
For Use With (Equipment) Pheos Safety Helmets
Material Elastic
Model 9790
Product Type Chin Strap