Få mere at vide
Greiner Bio-One 96-brønds skorstensdesignede mikroplader med rund (U) bund
Beskrivelse
- Non-pyrogenic
- Conical well design with round (U) bottom
- Ensures accurate pipetting
- Reduces dead volume of well
- Free of detectable DNase, RNase, human DNA
- Prevents wicking
Tekniske data
Tekniske data
| Format | Standard |
| Låg | Without Lid |
| Array | 8 x 12 |
| Farve | Neutral |
| Materiale | Polypropylene |
| Antal Wells | 96 |
| Sterilitet | Steril |
| Well Shape | Round Bottom (U-well) |
| Mængde | 100/Cs. |