missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Greiner Bio-One 96-well, Advanced TC Treated μClear™ F-Bottom/Chimney Well Microplates

Beskrivelse
Advanced TC polymer modification changes plastic surface in a way that increases primary and long-term cell adhesion and positively influences cell-dependant features and function enhanced cell attachment and higher proliferation rates facilitate rapid cell expansion even under restricted growth conditions.
- Provide conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield
- For use with serum reduced and serum-free media
- Allows differentiation of semi-adherent cells
- Non-pyrogenic and non-cytotoxic
- Homogenous cell growth
- Higher transgene activity
- μClear coated well bottom
Tekniske data
Tekniske data
| Udbrudt | No |
| Certificeringer/compliance | Free of detectable DNase/RNase, human DNA and pyrogens; Contents non-cytotoxic |
| Format | Standard |
| Låg | With Lid |
| Kulturområde | 34 mm2 |
| Array | 8 x 12 |
| Autoklaverbar | Not Autoclavable |
| Farve | Klar, Hvid, White |
| Materiale | Polystyrene |
| Antal Wells | 96 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?