Learn More
Uvex™ 9180 Futura Welding Goggles
Catalog Number 15715893
Click to view available options
Gray
Ergonomically designed, lightweight welding goggles with a very large field of vision and an excellent fit. Goggles feature a new filter technology with perfect color perception, and provide level 6 welding protection.
  • Ergonomic welding safety spectacles with wide field of vision and secure fit
  • Uvex duo-flex side arms with 4-stage side arm length adjustment and adjustable side arm inclination
  • Infradur coating is extremely scratch-resistant on both sides, while also providing protection against welding sparks
  • Goggles minimize damage caused by welding sparks
  • New filter technology ensures perfect color recognition
  • Level 6 welding protection
Lens Type Infradur
Description Welding Protection Level: 6
Certifications/Compliance (Europe) EN 166:2001, EN 169:2001
Lens Tint Gray
Frame Color Black, Green
Lens Material Polycarbonate
UV Protection UV 400