Learn More
Uvex™ 9180 Futura Welding Goggles
Click to view available options
Gray
Description
- Ergonomic welding safety spectacles with wide field of vision and secure fit
- Uvex duo-flex side arms with 4-stage side arm length adjustment and adjustable side arm inclination
- Infradur coating is extremely scratch-resistant on both sides, while also providing protection against welding sparks
- Goggles minimize damage caused by welding sparks
- New filter technology ensures perfect color recognition
- Level 6 welding protection
Specifications
Lens Type | Infradur |
Description | Welding Protection Level: 6 |
Certifications/Compliance (Europe) | EN 166:2001, EN 169:2001 |
Lens Tint | Gray |
Frame Color | Black, Green |
Lens Material | Polycarbonate |
UV Protection | UV 400 |