Få mere at vide
9-Oxo-10(9H)-acridineacetic Acid, TRC
Artikelnummer. 30136756
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136756 100 mg
30135612 500 mg
30003482 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136756 Leverandør Toronto Research Chemicals Leverandørnr. TRCO870098100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38609-97-1
Molekylær formel C15 H11 N O3
Molekylvægt (g/mol) 253.25
Synonym 10(9H)-Acridineacetic acid, 9-oxo-, 9-Oxo-10(9H)-acridineacetic acid, 10-(Carboxymethyl)-9-acridinone, 10-CMA, 9-Oxo-10-acridineacetic acid, Acridone-N-acetic acid, Cridanimod, L 1, N-(Carboxymethyl)-9-acridone, N-(Carboxymethyl)acridone
IUPAC navn 2-(9-oxoacridin-10-yl)acetic acid
SMIL OC(=O)CN1c2ccccc2C(=O)c3ccccc13
Tekniske data
Kemisk navn eller materiale Cridanimod
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C15H11NO3/c17-14(18)9-16-12-7-3-1-5-10(12)15(19)11-6-2-4-8-13(11)16/h1-8H,9H2,(H,17,18)
Formel vægt 253.0739
Procent renhed >95%