Få mere at vide
9-Nitroanthracene-d9, TRC
Artikelnummer. 30483646
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30483646 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483646 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4919310.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 9-Nitroanthracene-d9
CAS 220381-38-4
Renhedsgrad noter HPLC
Molekylær formel C14 D9 N O2
Mængde 0.5 mg
Synonym Anthracene-1,2,3,4,5,6,7,8,9-d9','10-nitro- (9CI), 10-Nitroanthracene-1,2,3,4,5,6,7,8,9-d9, 9-Nitroanthracene-1,2,3,4,5,6,7,8,10-d9, 9-Nitroanthracene-d9, 9-Nitroanthracene-D9
InChI formel InChI=1S/C14H9NO2/c16-15(17)14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9H/i1D,2D,3D,4D,5D,6D,7D,8D,9D
SMIL [2H]c1c([2H])c([2H])c2c(c3c([2H])c([2H])c([2H])c([2H])c3c([2H])c2c1[2H])[N+](=O)[O-]
IUPAC navn 1,2,3,4,5,6,7,8,9-nonadeuterio-10-nitroanthracene
Molekylvægt (g/mol) 232.282
Formel vægt 232.12
Procent renhed >95