Få mere at vide
9-Nitroanthracene-d9, CDN
Artikelnummer. 30320837
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320837 0.01 g
30313319 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320837 Leverandør CDN Leverandørnr. CDND43570.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 9-Nitroanthracene-d9
CAS 220381-38-4
Molekylær formel C14 D9 N O2
Mængde 0.01 g
Synonym Anthracene-1,2,3,4,5,6,7,8,9-d9, 10-nitro- (9 CI), 10-Nitroanthracene-1,2,3,4,5,6,7,8,9-d9, 9-Nitroanthracene-1,2,3,4,5,6,7,8,10-d9, 9-Nitroanthracene-d9, 9-Nitroanthracene-D9
InChI formel InChI=1 S/C14H9NO2/c16-15(17)14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D
SMIL [2 H]c1c([2 H])c([2 H])c2c(c3c([2 H])c([2 H])c([2 H])c([2 H])c3c([2 H])c2c1[2 H])[N+](=O)[O-]
IUPAC navn 1,2,3,4,5,6,7,8,9-nonadeuterio-10-nitroanthracene
Molekylvægt (g/mol) 232.282
Formel vægt 232.12 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity