Få mere at vide
9-Methylanthracene-d12, TRC
Artikelnummer. 30472428
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30472428 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472428 Leverandør Toronto Research Chemicals Leverandørnr. TRCM25891110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 9-Methylanthracene-d12
CAS 6406-97-9
Molekylær formel C15 D12
Mængde 10 mg
Synonym Anthracene-1,2,3,4,5,6,7,8,9-d9, 10-(methyl-d3)- (9CI, ACI), 10-(Methyl-d3)anthracene-1,2,3,4,5,6,7,8,9-d9 (ACI), Anthracene-d9, 9-(methyl-d3)- (7CI, 8CI), 9-Methylanthracene-d12, 9-Methylanthracene-D12
InChI formel InChI=1S/C15H12/c1-11-14-8-4-2-6-12(14)10-13-7-3-5-9-15(11)13/h2-10H,1H3/i1D3,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c([2H])c2c(c3c([2H])c([2H])c([2H])c([2H])c3c([2H])c2c1[2H])C([2H])([2H])[2H]
IUPAC navn 1,2,3,4,5,6,7,8,9-nonadeuterio-10-(trideuteriomethyl)anthracene
Molekylvægt (g/mol) 204.33
Formel vægt 204.169