Få mere at vide
9-Methyl Adenine-d3, TRC
Artikelnummer. 30473574
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30473574 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473574 Leverandør Toronto Research Chemicals Leverandørnr. TRCM27591510MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 9-Methyl-d3-adenine
CAS 130859-46-0
Molekylær formel C6 2H3 H4 N5
Mængde 10 mg
Synonym 9-Methyl Adenine D3
InChI formel InChI=1S/C6H7N5/c1-11-3-10-4-5(7)8-2-9-6(4)11/h2-3H,1H3,(H2,7,8,9)/i1D3
SMIL [2H]C([2H])([2H])n1cnc2c(N)ncnc12
IUPAC navn 9-(trideuteriomethyl)purin-6-amine
Molekylvægt (g/mol) 152.17
Formel vægt 152.089