Få mere at vide
9-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one, Mikromol™
Artikelnummer. 30050637
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30050637 50 mg
1 options
Artikelnummer. 30050637 Leverandør Mikromol Leverandørnr. MM0095.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
9-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 9-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]-benzimidazol-2(12H)-one
CAS 125656-83-9
Analyt- eller komponentnavne 9-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]-benzimidazol-2(12H)-one
Forsendelsestilstand Room Temperature
Molekylær formel C16 H13 N3 O2 S
Certificeringer/compliance ISO 17025
Synonym Omeprazole Imp. G (EP), Omeprazole USP Related Compound F and G Mixture, Omeprazole USP RC F and G Mixture, 9-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one, Omeprazole Related Compound G, Esomeprazole Sodium Impurity G, Omeprazole Impurity G, Esomeprazole Impurity G
InChI formel InChI=1S/C16H13N3O2S/c1-8-7-18-13(9(2)14(8)20)15(22)19-12-6-10(21-3)4-5-11(12)17-16(18)19/h4-7H,1-3H3
SMIL COc1ccc2nc3N4C=C(C)C(=O)C(=C4C(=S)n3c2c1)C
IUPAC navn 9-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
Molekylvægt (g/mol) 311.36
Mængde 50 mg
Formel vægt 311.0728 g/mol