Få mere at vide
9-Hydroxyellipticine Hydrochloride, TRC
Artikelnummer. 17973205
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17973205 50 mg
17973405 10 mg
17923515 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17973205 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94968350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 52238-35-4
Molekylær formel C17 H14 N2 O . H Cl
Molekylvægt (g/mol) 298.767
Synonym 5,11-Dimethyl-6H-pyrido[4,3-b]carbazol-9-ol Hydrochloride;
IUPAC navn 5,11-dimethyl-6H-pyrido[4,3-b]carbazol-9-ol;hydrochloride
SMIL Cl.Cc1c2ccncc2c(C)c3c4cc(O)ccc4[nH]c13
Tekniske data
Kemisk navn eller materiale 9-Hydroxyellipticine Hydrochloride
Mængde 50 mg
InChI formel InChI=1S/C17H14N2O.ClH/c1-9-14-8-18-6-5-12(14)10(2)17-16(9)13-7-11(20)3-4-15(13)19-17;/h3-8,19-20H,1-2H3;1H
Formel vægt 298.087