Få mere at vide
9-epi-Artemisinin, TRC
Artikelnummer. 30137270
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137270 1 g
30010653 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137270 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7775051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 113472-97-2
Molekylær formel C15H22O5
Molekylvægt (g/mol) 282.33
Synonym (3R,5aS,6R,8aS,9S,12S,12aR)-Octahydro-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin-10(3H)-one, (+)-9-Epiartemisinin, 11-epi-Artemisinin;
IUPAC navn (1R,4S,5R,8S,9S,12S,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.04,13.08,13]hexadecan-10-one
SMIL C[C@@H]1CC[C@H]2[C@@H](C(=O)O[C@H]3[C@@]24[C@H]1CC[C@](O3)(OO4)C)C
Tekniske data
Kemisk navn eller materiale 9-epi-Artemisinin
Analyt- eller komponentnavne 9-epi-Artemisinin
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C15H22O5/c1-8-4-5-11-9(2)12(16)17-13-15(11)10(8)6-7-14(3,18-13)19-20-15/h8-11,13H,4-7H2,1-3H3/t8-,9+,10+,11+,13-,14-,15-/m1/s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 1 g
Formel vægt 282.1467
Procent renhed >95%