Learn More
9-Deazaguanosine, TRC
Product Code. 30357007
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30357007 10 mg 10mg
1 options
Product Code. 30357007 Supplier Toronto Research Chemicals Supplier No. TRCD20140010MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 9-Deazaguanosine
CAS 102731-45-3
Molecular Formula C11 H14 N4 O5
Quantity 10 mg
InChI Formula InChI=1S/C11H14N4O5/c12-11-13-8-4(9(19)14-11)1-2-15(8)10-7(18)6(17)5(3-16)20-10/h1-2,5-7,10,16-18H,3H2,(H3,12,13,14,19)/t5-,6-,7-,10-/m1/s1
SMILES NC1=Nc2c(ccn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1
IUPAC Name 2-amino-7-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-pyrrolo[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol) 282.25
Formula Weight 282.0964