missing translation for 'onlineSavingsMsg'
Learn More

9-cis-retinsyre, TRC

Product Code. 30134331
Change view
Click to view available options
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30134331 5 mg
18081220 10 mg
18031720 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134331 Supplier Toronto Research Chemicals Supplier No. TRCR2450005MG
 Begrænset vare
Forventet forsendelse: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5300-03-8
Molekylær formel C20 H28 O2
Molekylvægt (g/mol) 300.44
Synonym 9-cis-Retinoic Acid, Retinoic acid, cis-9,trans-13- (8CI), 9-cis-Retinoic acid, 9(Z)-Retinoic acid, 9-cis-Tretinoin, AGN 192013, ALRT 1057, Alitretinoin, LG 100057, LGD 100057, LGD 1057, NSC 659772, Panretin, Panretyn, Panrexin, Toctino, BAL0004079-000, code for free acid, BAL4079, 9-cis retinoic acid, 9-(Z) Retinoic Acid, R0004079-000, CRA-Alitretinoin, INN Alitretinoin
IUPAC navn (2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenosyre
SMIL C\C(=C\C=C\C(=C\C(=O)O)\C)\C=C\C1=C(C)CCCC1(C)C

Specifications

Kemisk navn eller materiale 9-cis-Retinoic acid
Mængde 5 mg
InChI formel InChI=1S/C20H28O2/c1-15(8-6-9-16(2)14-19(21)22)11-12-18-17(3)10-7-13-20(18,4)5/h6,8-9,11-12,14H,7,10,13H2,1-5H3,(H,21,22)/b9-6+,12-11+,15-8-,16-14+
Formel vægt 300.2089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.