missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
9-cis-5,6-Epoxy-5,6-dihydro-retinoic Acid, TRC
Specifications
Specifications
| Kemisk navn eller materiale | 9-cis-5,6-Epoxy-5,6-dihydroretinoic Acid (5,6-Epoxialitretinoine) |
| CAS | 1431303-55-7 |
| Molekylær formel | C20 H28 O3 |
| Mængde | 1 mg |
| Synonym | 9-cis-5,6-Epoxy-5,6-dihydro-retinoic Acid, Retinoic acid, 5,6-epoxy-5,6-dihydro-, 9-cis-, BAL19222, GSK3040320A, BAL0019222-000, BAL0004079-000, BAL4079, 9-cis retinoic acid, alitretinoin related |
| InChI formel | InChI=1S/C20H28O3/c1-15(8-6-9-16(2)14-17(21)22)10-13-20-18(3,4)11-7-12-19(20,5)23-20/h6,8-10,13-14H,7,11-12H2,1-5H3,(H,21,22)/b9-6+,13-10+,15-8-,16-14+ |
| SMIL | C\C(=C\C=C\C(=C\C(=O)O)\C)\C=C\C12OC1(C)CCCC2(C)C |
| IUPAC navn | (2E,4E,6Z,8E)-3,7-dimethyl-9-(1,5,5-trimethyl-7-oxabicyclo[4.1.0]heptan-6-yl)nona-2,4,6,8-tetraenoic acid |
| Molekylvægt (g/mol) | 316.435 |
| Formel vægt | 316.204 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?